Order Entry
Export
ContactUsLinkComponent
2,2-Bis(1,3,5-trimethylphenyl)-1,1,1-trifluoroethane
Irritant
2,2-Bis(1,3,5-trimethylphenyl)-1,1,1-trifluoroethane
Catalog # APOSPC450067-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,2-Bis(1,3,5-trimethylphenyl)-1,1,1-trifluoroethane
Catalog # APOSPC450067-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:115…119 °C
 :320.392 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1314534-82-1
Formula:C₂₀H₂₃F₃
MDL Number:MFCD22123955