Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2,2-difluorohex-3-enoate
Ethyl 2,2-difluorohex-3-enoate
Ethyl 2,2-difluorohex-3-enoate
Ethyl 2,2-difluorohex-3-enoate
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl 2,2-difluorohex-3-enoate
Supplier:
Apollo Scientific
About this item
CAS Number:
1346523-54-3
:
Formula:
C₈H₁₂F₂O₂
MDL Number:
MFCD23704335
:
178.176 g/mol
Storage Temperature:
Ambient