4,6-Bis(trifluoromethyl)-2(1H)-pyrimidinethione
Catalog # APOSPC450036-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:248.149 g/mol
Storage Temperature:Ambient
CAS Number:402520-53-0
Formula:C₆H₂F₆N₂S
MDL Number:MFCD26408189