Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl 2,2-difluoro-2-(3,4-dichlorophenyl)acetate
Supplier:
Apollo Scientific
About this item
CAS Number:
56177-76-5
:
Formula:
C₁₀H₈Cl₂F₂O₂
MDL Number:
MFCD01937334
:
269.072 g/mol
Storage Temperature:
Ambient