1,2-Dibromofluoroethylene
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:358-98-5
- Formula:C₂HBr₂F
- MDL Number:MFCD02913516
- Molecular Weight:203.836 g/mol
- Boiling Pt:72 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:358-98-5
- Formula:C₂HBr₂F
- MDL Number:MFCD02913516
- Molecular Weight:203.836 g/mol
- Boiling Pt:72 °C
- Storage Temperature:Ambient



