Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Dibromo-2-fluoroethylene
1,1-Dibromo-2-fluoroethylene 97%
1,1-Dibromo-2-fluoroethylene 97%
1,1-Dibromo-2-fluoroethylene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1,1-Dibromo-2-fluoroethylene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
358-96-3
:
Formula:
C₂HBr₂F
MDL Number:
MFCD02913515
:
203.836 g/mol
Boiling Pt:
90…91 °C
Storage Temperature:
Ambient