3-(3,5-Dichlorophenylcarbamoyl)-2-fluorobenzoic acid
Catalog # APOSPC430595-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:328.123 g/mol
Storage Temperature:Ambient
CAS Number:1206969-96-1
Formula:C₁₄H₈Cl₂FNO₃
MDL Number:MFCD14585330