2,3,3-Trifluoro-1,4-benzodioxen-5-ol
Catalog # APOSPC430558-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:206.119 g/mol
Storage Temperature:Ambient
CAS Number:1008452-12-7
Formula:C₈H₅F₃O₃
MDL Number:MFCD04972980