4,5-Difluoroanthranlic acid
Catalog # APOSPC430548-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:258.22 g/mol
Storage Temperature:Ambient
CAS Number:1258652-67-3
Formula:C₁₅H₈F₂O₂
MDL Number:MFCD18157739