Ethyl 2,2-difluoro-3-hydroxy-3-phenylpropanoate
Catalog # APOSPC430249-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.208 g/mol
Storage Temperature:Ambient
CAS Number:92207-60-8
Formula:C₁₁H₁₂F₂O₃
MDL Number:MFCD00725033