Order Entry
Export
ContactUsLinkComponent
Methyl 3-fluoroisonicotinate
Methyl 3-fluoroisonicotinate
Catalog # APOSPC430141-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-fluoroisonicotinate
Catalog # APOSPC430141-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.126 g/mol
Storage Temperature:Ambient
CAS Number:876919-08-3
Formula:C₇H₆FNO₂
MDL Number:MFCD11045048