Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-6-iodoaniline
2,4-Difluoro-6-iodoaniline
2,4-Difluoro-6-iodoaniline
Toxic
Irritant
2,4-Difluoro-6-iodoaniline
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
Irritant
2,4-Difluoro-6-iodoaniline
Supplier:
Apollo Scientific
About this item
CAS Number:
582319-15-1
:
Formula:
C₆H₄F₂IN
MDL Number:
MFCD08059536
:
255.004 g/mol
UN:
2811
ADR:
6.1,III
:
GHS06
:
GHS07
Storage Temperature:
Ambient