
2,6-Difluoro-4-nitroanisole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:392-25-6
:
Formula:C₇H₅F₂NO₃
MDL Number:MFCD06656243
:189.116 g/mol
Boiling Pt:71 °C (0.6 mm Hg)
Melting Pt:33…37 °C
:GHS07
Storage Temperature:Ambient