Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(3,3-Difluorocyclobutyl)formamide
N-(3,3-Difluorocyclobutyl)formamide 98%
N-(3,3-Difluorocyclobutyl)formamide 98%
N-(3,3-Difluorocyclobutyl)formamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
N-(3,3-Difluorocyclobutyl)formamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1355328-30-1
:
Formula:
C₅H₇F₂NO
MDL Number:
MFCD24619684
:
135.112 g/mol
Storage Temperature:
Ambient