Order Entry
Export
ContactUsLinkComponent
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-3-ol 95%
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-3-ol 95%
Catalog # APOSPC421210-100MG
Supplier:  Apollo Scientific
CAS Number:  
3-(Trifluoromethyl)-2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-3-ol 95%
Catalog # APOSPC421210-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :204.149 g/mol
Storage Temperature:Ambient
CAS Number:892414-46-9
Formula:C₈H₇F₃N₂O
MDL Number:MFCD24556540