Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-(difluoromethyl)-1,3-thiazole
5-Bromo-2-(difluoromethyl)-1,3-thiazole 95%
5-Bromo-2-(difluoromethyl)-1,3-thiazole 95%
Irritant
5-Bromo-2-(difluoromethyl)-1,3-thiazole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Bromo-2-(difluoromethyl)-1,3-thiazole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1319255-36-1
:
Formula:
C₄H₂BrF₂NS
MDL Number:
MFCD23106373
:
214.031 g/mol
:
GHS07
Storage Temperature:
Freezer