Order Entry
Export
ContactUsLinkComponent
2,4-Difluoro-1-(2-fluorophenyl)benzene 95%
2,4-Difluoro-1-(2-fluorophenyl)benzene 95%
Catalog # APOSPC421049-250MG
Supplier:  Apollo Scientific
CAS Number:  
2,4-Difluoro-1-(2-fluorophenyl)benzene 95%
Catalog # APOSPC421049-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :208.179 g/mol
Storage Temperature:Ambient
CAS Number:115245-05-1
Formula:C₁₂H₇F₃
MDL Number:MFCD22051120