2-(3,4-Difluorophenyl)-4'-fluoroacetophenone
Catalog # APOSPC421006-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.216 g/mol
Storage Temperature:Ambient
CAS Number:1507249-35-5
Formula:C₁₄H₉F₃O
MDL Number:MFCD24968600