Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-1,1-difluorocyclobutane
3-Bromo-1,1-difluorocyclobutane
3-Bromo-1,1-difluorocyclobutane
Flammable
Irritant
3-Bromo-1,1-difluorocyclobutane
Supplier:
Apollo Scientific
Certificates
Expand image
Flammable
Irritant
3-Bromo-1,1-difluorocyclobutane
Supplier:
Apollo Scientific
About this item
CAS Number:
1310729-91-9
:
Formula:
C₄H₅BrF₂
MDL Number:
MFCD22989493
:
170.983 g/mol
UN:
1993
ADR:
3,III
:
GHS02
:
GHS07
Storage Temperature:
Refrigerator