Methyl 3-(3,3-difluorocyclobutyl)-3-oxopropanoate
Catalog # APOSPC420002-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.16 g/mol
Storage Temperature:Ambient
CAS Number:1191096-28-2
Formula:C₈H₁₀F₂O₃
MDL Number:MFCD17015923