Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2,4-difluoro-6-methylpyrimidine
5-Chloro-2,4-difluoro-6-methylpyrimidine 98%
5-Chloro-2,4-difluoro-6-methylpyrimidine 98%
5-Chloro-2,4-difluoro-6-methylpyrimidine 98%
APOSPC4191-1G
:
Apollo Scientific
:
5-Chloro-2,4-difluoro-6-methylpyrimidine 98%
APOSPC4191-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
164.541 g/mol
:
GHS07
:
Ambient
:
72630-78-5
:
C₅H₃ClF₂N₂
:
MFCD00036058