Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-2-methoxyphenylboronic acid
3,4-Difluoro-2-methoxyphenylboronic acid 98%
3,4-Difluoro-2-methoxyphenylboronic acid 98%
3,4-Difluoro-2-methoxyphenylboronic acid 98%
APOSPC412110-250MG
:
Apollo Scientific
:
3,4-Difluoro-2-methoxyphenylboronic acid 98%
APOSPC412110-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
187.936 g/mol
:
Ambient
:
905583-06-4
:
C₇H₇BF₂O₃
:
MFCD13181640