Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3,3-Difluorocyclobutyl)ethan-1-one
1-(3,3-Difluorocyclobutyl)ethan-1-one
1-(3,3-Difluorocyclobutyl)ethan-1-one
1-(3,3-Difluorocyclobutyl)ethan-1-one
Supplier:
Apollo Scientific
Certificates
Expand image
1-(3,3-Difluorocyclobutyl)ethan-1-one
Supplier:
Apollo Scientific
About this item
CAS Number:
1621223-57-1
:
Formula:
C₆H₈F₂O
MDL Number:
MFCD28501817
:
134.124 g/mol
Storage Temperature:
Ambient