(2,2-Difluorocyclobutyl)methanamine hydrochloride
Catalog # APOSPC411269-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.589 g/mol
Storage Temperature:Ambient
CAS Number:2055839-95-5
Formula:C₅H₉F₂N·ClH
MDL Number:MFCD30530422