2-(2,2,2-Trifluoroethoxy)pyridin-3-amine hydrochloride
Catalog # APOSPC410106-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.599 g/mol
Storage Temperature:Ambient
CAS Number:1431964-68-9
Formula:C₇H₇F₃N₂O·ClH
MDL Number:MFCD25368944