Order Entry
Export
ContactUsLinkComponent
1-(2,2-Difluoroethyl)-3-nitro-1H-1,2,4-triazole
1-(2,2-Difluoroethyl)-3-nitro-1H-1,2,4-triazole
Catalog # APOSPC410074-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,2-Difluoroethyl)-3-nitro-1H-1,2,4-triazole
Catalog # APOSPC410074-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.097 g/mol
Storage Temperature:Ambient
Formula:C₄H₄F₂N₄O₂
MDL Number:MFCD09701856