Order Entry
Export
ContactUsLinkComponent
6-(4-Fluorophenyl)-3-methyl[1,2]oxazolo[5,4-b]pyridine-4-carbohydrazide
6-(4-Fluorophenyl)-3-methyl[1,2]oxazolo[5,4-b]pyridine-4-carbohydrazide
Catalog # APOSPC410042-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(4-Fluorophenyl)-3-methyl[1,2]oxazolo[5,4-b]pyridine-4-carbohydrazide
Catalog # APOSPC410042-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.261 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₁FN₄O₂
MDL Number:MFCD08556240