Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3,3-Difluorocyclobutyl)ethanol
(3,3-Difluorocyclobutyl)ethanol
(3,3-Difluorocyclobutyl)ethanol
(3,3-Difluorocyclobutyl)ethanol
APOSPC409042-1G
:
Apollo Scientific
:
(3,3-Difluorocyclobutyl)ethanol
APOSPC409042-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
136.14 g/mol
:
Ambient
:
1987
:
3,III
:
GHS02
:
GHS07
:
1056467-54-9
:
C₆H₁₀F₂O
:
MFCD18790781