Order Entry
Export
ContactUsLinkComponent
5,6-Difluoro-2,3-dihydro-1H-inden-1-one 97%
5,6-Difluoro-2,3-dihydro-1H-inden-1-one 97%
Catalog # APOSPC408504-1G
Supplier:  Apollo Scientific
CAS Number:  
5,6-Difluoro-2,3-dihydro-1H-inden-1-one 97%
Catalog # APOSPC408504-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.14 g/mol
Storage Temperature:Ambient
CAS Number:161712-77-2
Formula:C₉H₆F₂O
MDL Number:MFCD07772125