N-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Catalog # APOSPC408352-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:258.763 g/mol
Storage Temperature:Ambient
CAS Number:1353946-54-9
Formula:C₁₃H₁₉FN₂·ClH
MDL Number:MFCD21098651