N,N'-Bis-(3-fluoro-benzyl)-cyclohexane-1,4-diamine
Catalog # APOSPC408339-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:330.415 g/mol
Storage Temperature:Ambient
CAS Number:1353987-21-9
Formula:C₂₀H₂₄F₂N₂
MDL Number:MFCD21098468