
1,4-Bis(bromomethyl)tetrafluorobenzene
Catalog # APOSPC408158-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Melting Pt:88…90 °C
:335.919 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:776-40-9
Formula:C₈H₄Br₂F₄
MDL Number:MFCD07779350