Order Entry
Export
ContactUsLinkComponent
5-Fluoro-1,2-dihydroisoquinolin-1-one ≥98%
5-Fluoro-1,2-dihydroisoquinolin-1-one ≥98%
Catalog # APOSPC407077-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Fluoro-1,2-dihydroisoquinolin-1-one ≥98%
Catalog # APOSPC407077-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :163.148 g/mol
Storage Temperature:Refrigerator
CAS Number:410086-25-8
Formula:C₉H₆FNO
MDL Number:MFCD12405865