
2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate
Catalog # APOSPC405006-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:255.191 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1523572-02-2
Formula:C₇H₁₁NO₂·CF₃CO₂H
MDL Number:MFCD19982773