3,4-Dibromo-6,8-difluoroquinoline
Catalog # APOSPC404607-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:322.932 g/mol
Storage Temperature:Ambient
CAS Number:1210168-52-7
Formula:C₉H₃Br₂F₂N
MDL Number:MFCD13619737