Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dichloro-5,8-difluoroquinoline
3,4-Dichloro-5,8-difluoroquinoline
3,4-Dichloro-5,8-difluoroquinoline
3,4-Dichloro-5,8-difluoroquinoline
APOSPC404601-1G
:
Apollo Scientific
:
3,4-Dichloro-5,8-difluoroquinoline
APOSPC404601-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
234.03 g/mol
:
Ambient
:
1204811-35-7
:
C₉H₃Cl₂F₂N
:
MFCD13192997