
3,4-Dichloro-8-fluoroquinoline
Catalog # APOSPC404598-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.039 g/mol
Melting Pt:107…108 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1204811-28-8
Formula:C₉H₄Cl₂FN
MDL Number:MFCD13192963