3,4-Dichloro-7-fluoroquinoline
Catalog # APOSPC404597-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.039 g/mol
Storage Temperature:Ambient
CAS Number:1204810-51-4
Formula:C₉H₄Cl₂FN
MDL Number:MFCD13192961