
3,4-Dichloro-6-fluoroquinoline
Catalog # APOSPC404595-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.039 g/mol
Melting Pt:80…81 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1204810-46-7
Formula:C₉H₄Cl₂FN
MDL Number:MFCD13192959