4-Chloro-5,7-difluoro-2-methylquinoline
Catalog # APOSPC404588-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.611 g/mol
Storage Temperature:Ambient
CAS Number:288151-41-7
Formula:C₁₀H₆ClF₂N
MDL Number:MFCD11501279