4-Bromo-5,7-difluoroquinoline
Catalog # APOSPC404581-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.036 g/mol
Storage Temperature:Ambient
CAS Number:1189107-48-9
Formula:C₉H₄BrF₂N
MDL Number:MFCD12674965