Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,8-Bis(trifluoromethyl)-4-vinylquinoline
2,8-Bis(trifluoromethyl)-4-vinylquinoline
2,8-Bis(trifluoromethyl)-4-vinylquinoline
2,8-Bis(trifluoromethyl)-4-vinylquinoline
APOSPC404566-1G
:
Apollo Scientific
:
2,8-Bis(trifluoromethyl)-4-vinylquinoline
APOSPC404566-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
291.192 g/mol
:
GHS07
:
Ambient
:
1031928-53-6
:
C₁₃H₇F₆N
:
MFCD10703428