6-Fluoro-2-hydroxy-3-phenylquinoline
Catalog # APOSPC404565-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:239.244 g/mol
Storage Temperature:Ambient
CAS Number:1031928-37-6
Formula:C₁₅H₁₀FNO
MDL Number:MFCD09997959