Order Entry
Export
ContactUsLinkComponent
2-[2,8-Bis(trifluoromethyl)-4-quinolyl]oxirane
2-[2,8-Bis(trifluoromethyl)-4-quinolyl]oxirane
Catalog # APOSPC404553-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2,8-Bis(trifluoromethyl)-4-quinolyl]oxirane
Catalog # APOSPC404553-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :307.191 g/mol
Storage Temperature:Ambient
CAS Number:57120-54-4
Formula:C₁₃H₇F₆NO
MDL Number:MFCD09260425