Order Entry
Export
ContactUsLinkComponent
3,5-Difluoropyridine-2-sulphonyl chloride
3,5-Difluoropyridine-2-sulphonyl chloride
Catalog # APOSPC400562-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Difluoropyridine-2-sulphonyl chloride
Catalog # APOSPC400562-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :213.59 g/mol
Storage Temperature:Ambient
CAS Number:1261587-79-4
Formula:C₅H₂ClF₂NO₂S
MDL Number:MFCD18411876