4-Fluoro-1,4'-bipiperidine hydrochloride
Catalog # APOSPC400530-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:222.731 g/mol
Storage Temperature:Ambient
CAS Number:1426290-04-1
Formula:C₁₀H₁₉FN₂·ClH
MDL Number:MFCD23097075