Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-5-fluoropyridine-3,4-diamine
2-Bromo-5-fluoropyridine-3,4-diamine
2-Bromo-5-fluoropyridine-3,4-diamine
2-Bromo-5-fluoropyridine-3,4-diamine
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-5-fluoropyridine-3,4-diamine
Supplier:
Apollo Scientific
About this item
CAS Number:
1227958-29-3
:
Formula:
C₅H₅BrFN₃
MDL Number:
MFCD19443191
:
206.016 g/mol
Storage Temperature:
Ambient