
1-(Azetidin-3-yl)-4,4-difluoropiperidine
Catalog # APOSPC400503-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:176.207 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1257293-83-6
Formula:C₈H₁₄F₂N₂
MDL Number:MFCD22123272