Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-4,4'-Difluorochalcone
trans-4,4'-Difluorochalcone 98%
trans-4,4'-Difluorochalcone 98%
:
Apollo Scientific
trans-4,4'-Difluorochalcone 98%
:
Apollo Scientific
:
102692-35-3
:
:
C₁₅H₁₀F₂O
:
MFCD00219906
:
117…119 °C
:
244.236 g/mol
:
GHS07
:
Ambient