Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Trifluoromethyl)-1,3-benzothiazol-2-ylamine
Supplier:
Apollo Scientific
About this item
CAS Number:
60388-36-5
:
Formula:
C₈H₅F₃N₂S
MDL Number:
MFCD00160038
:
218.199 g/mol
Storage Temperature:
Ambient